Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 8 | 829 | ||
XM_005263600.2 | XP_005263657.1 | 8 | 829 | |||
XM_005263603.2 | XP_005263660.1 | 9 | 863 | |||
XM_005263605.3 | XP_005263662.1 | 9 | 864 | |||
XM_011510667.1 | XP_011508969.1 | 8 | 829 | |||
XM_011510668.2 | XP_011508970.1 | 5 | 389 | |||
XM_011510669.2 | XP_011508971.1 | 9 | 873 | |||
XM_017003407.1 | XP_016858896.1 | 8 | 829 | |||
XM_017003408.1 | XP_016858897.1 | 10 | 993 | |||
GenBank mRNA | AF452722.1 | - | 8 | 685 | ||
AK290072.1 | - | 8 | 829 | |||
AK303653.1 | - | 9 | 868 | |||
AY343910.1 | - | 8 | 714 | |||
BC120992.1 | - | 8 | 700 | 40116411 |