Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SMYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198274.3 | NP_938015.1 | 7 | 1022 | ||
XM_005264156.2 | XP_005264213.1 | 6 | 938 | |||
GenBank mRNA | AK091724.1 | - | 7 | 940 | ||
AL832035.1 | - | 7 | 939 | |||
AY518933.1 | - | 7 | 919 | |||
AY518934.1 | - | 3 | 288 | |||
BC126191.1 | - | 7 | 1022 | 8991907 | ||
BX647878.1 | - | 7 | 941 |