Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT10 SIRNA Assays ›
Gene Symbol:
SEPT10
Gene Name:
septin 10
Gene Aliases:
-
Chromosome Location:
Chr.2: 109542797 - 109614247 on Build GRCh38
UniGene ID:
Hs.469615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45506
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321496.1
NP_001308425.1
3
574
NM_001321498.1
NP_001308427.1
5
888
NM_001321500.1
NP_001308429.1
4
819
NM_001321501.1
NP_001308430.1
5
892
NM_001321502.1
NP_001308431.1
4
819
NM_001321504.1
NP_001308433.1
2
505
NM_001321505.1
NP_001308434.1
2
505
NM_001321507.1
NP_001308436.1
3
574
NM_001321508.1
NP_001308437.1
2
505
NM_001321509.1
NP_001308438.1
5
892
NM_001321512.1
NP_001308441.1
5
888
NM_001321513.1
NP_001308442.1
5
1025
NM_001321514.1
NP_001308443.1
5
716
NM_001321515.1
NP_001308444.1
4
956
NM_144710.4
NP_653311.1
5
888
NM_178584.3
NP_848699.1
4
819
XM_006712317.1
XP_006712380.1
5
888
XM_011510698.2
XP_011509000.1
5
721
XM_011510699.2
XP_011509001.1
4
540
XM_011510700.2
XP_011509002.1
5
891
XM_011510701.2
XP_011509003.1
3
615
XM_011510703.2
XP_011509005.1
5
498
XM_011510704.2
XP_011509006.1
5
888
GenBank mRNA
AF146760.1
-
5
635
AK021681.1
-
5
888
AK022682.1
-
2
220
AK092033.1
-
4
539
AK292038.1
-
5
606
AK295560.1
-
3
284
BC020502.1
-
5
582
3913356
BC050345.1
-
5
825
4830029
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
cell division
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0