Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207330.2 | NP_997213.1 | 2 | 160 | ||
XM_017007784.1 | XP_016863273.1 | 2 | 117 | |||
GenBank mRNA | AK095299.1 | - | 2 | 119 | ||
BC067881.1 | - | 2 | 158 | 30345943 | ||
CB960388.1 | - | 2 | 153 | |||
CR749484.1 | - | 2 | 159 |