Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 1 | 39 | |||
GenBank mRNA | DC319181.1 | - | 1 | 42 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 1 | 39 | ||
GenBank mRNA | AK289708.1 | - | 1 | 42 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 1 | 227 | |||
GenBank mRNA | AK056232.1 | - | 1 | 78 |