Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005198.4 | NP_005189.2 | 10 | 1317 | ||
GenBank mRNA | AB029886.1 | - | 10 | 1167 | ||
AK097775.1 | - | 9 | 1652 | |||
AK314324.1 | - | 10 | 1151 | |||
AL096780.1 | - | 10 | 1242 | |||
AL096781.1 | - | 10 | 1099 | |||
BC024286.1 | - | 10 | 1096 | |||
BC082263.1 | - | 10 | 1154 | 6384695 | ||
BC101488.1 | - | 10 | 1162 | 8068994 | ||
BC113521.1 | - | 10 | 1162 | 8322573 | ||
CR456419.1 | - | 10 | 1228 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 10 | 1317 | |||
GenBank mRNA | AB051457.1 | - | 7 | 1955 | ||
AK301477.1 | - | 8 | 773 | |||
BC131570.1 | - | 1 | 31 | 40108355 |