Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRIQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105659.1 | NP_001099129.1 | 7 | 1235 | ||
NM_001322315.1 | NP_001309244.1 | 7 | 1235 | |||
XM_011540658.1 | XP_011538960.1 | 6 | 925 | |||
XM_011540659.1 | XP_011538961.1 | 5 | 439 | |||
XM_011540660.1 | XP_011538962.1 | 5 | 2564 | |||
XM_011540662.2 | XP_011538964.1 | 4 | 863 | |||
XM_011540663.2 | XP_011538965.1 | 6 | 1119 | |||
GenBank mRNA | AK131476.2 | - | 9 | 3557 |