Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 7 | 1117 | ||
XM_006710360.3 | XP_006710423.2 | 4 | 392 | |||
XM_011540683.2 | XP_011538985.1 | 7 | 1079 | |||
XM_011540684.2 | XP_011538986.1 | 7 | 1204 | |||
XM_011540685.2 | XP_011538987.1 | 7 | 1216 | |||
XM_011540688.2 | XP_011538990.1 | 7 | 1079 | |||
XM_011540690.2 | XP_011538992.1 | 7 | 1079 | |||
XM_011540691.2 | XP_011538993.1 | 7 | 1079 | |||
XM_011540692.2 | XP_011538994.1 | 7 | 1073 | |||
XM_011540693.1 | XP_011538995.1 | 7 | 1079 | |||
XM_011540695.1 | XP_011538997.1 | 7 | 1079 | |||
XM_011540698.1 | XP_011539000.1 | 7 | 1079 | |||
GenBank mRNA | AK125833.1 | - | 7 | 1265 | ||
AK295797.1 | - | 7 | 1101 |