Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMZP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012230.3 | NP_036362.3 | 1 | 533 | ||
NM_152992.2 | NP_694537.1 | 1 | 533 | |||
GenBank mRNA | U10099.1 | - | 1 | 479 |
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB354586.1 | - | 1 | 512 |