Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016379.3 | NP_057463.2 | 1 | 88 | ||
GenBank mRNA | AF167078.1 | - | 1 | 88 |