Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271862.1 | NP_001258791.1 | 5, 6 | 436 | ||
NM_173516.2 | NP_775787.1 | 5, 6 | 535 | |||
XM_011535491.2 | XP_011533793.1 | 5, 6 | 462 | |||
XM_017010312.1 | XP_016865801.1 | 5, 6 | 1224 | |||
XM_017010313.1 | XP_016865802.1 | 3, 4 | 1110 | |||
XM_017010314.1 | XP_016865803.1 | 5, 6 | 1224 | |||
XM_017010315.1 | XP_016865804.1 | 3, 4 | 314 | |||
XM_017010316.1 | XP_016865805.1 | 3, 4 | 379 | |||
GenBank mRNA | AK093139.1 | - | 5, 6 | 413 | ||
AK097559.1 | - | 5, 6 | 515 | |||
BC112246.1 | - | 5, 6 | 419 | 8327714 |