Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003520.3 | NP_003511.1 | 1 | 60 | ||
GenBank mRNA | AK312228.1 | - | 1 | 169 | ||
BC101411.1 | - | 1 | 60 | 40021691 | ||
BC101412.1 | - | 1 | 60 | 40021693 | ||
BC101413.1 | - | 1 | 60 | 40021695 |