Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM66 SIRNA Assays ›
Gene Symbol:
TRIM66
Gene Name:
tripartite motif containing 66
Gene Aliases:
C11orf29, TIF1D, TIF1DELTA
Chromosome Location:
Chr.11: 8612037 - 8683217 on Build GRCh38
UniGene ID:
Hs.130836
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s458620
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014818.1
NP_055633.1
20
5550
XM_006718397.2
XP_006718460.1
25
6488
XM_006718398.3
XP_006718461.1
21
7100
XM_011520504.1
XP_011518806.1
26
6531
XM_011520507.2
XP_011518809.1
25
6452
XM_011520508.1
XP_011518810.1
25
6531
XM_011520509.1
XP_011518811.1
24
6413
XM_011520510.1
XP_011518812.1
25
6404
XM_011520511.1
XP_011518813.1
25
6377
XM_011520512.1
XP_011518814.1
21
5752
XM_011520513.1
XP_011518815.1
21
5750
XM_011520514.2
XP_011518816.1
21
5706
XM_011520515.1
XP_011518817.1
22
5919
XM_011520516.1
XP_011518818.1
22
5919
XM_011520517.2
XP_011518819.1
22
7241
XM_011520518.1
XP_011518820.1
22
5889
XM_011520519.1
XP_011518821.1
21
5777
XM_011520522.2
XP_011518824.1
20
6955
XM_011520523.2
XP_011518825.1
21
7837
XM_011520526.2
XP_011518828.1
13
4806
XM_011520527.1
XP_011518829.1
13
4074
XM_017018629.1
XP_016874118.1
25
6881
XM_017018630.1
XP_016874119.1
20
5843
XM_017018631.1
XP_016874120.1
20
5995
GenBank mRNA
AB002296.2
-
19
5804
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0