Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024607.2 | NP_001019778.1 | 2 | 265 | ||
GenBank mRNA | BG211856.1 | - | 2 | 266 |