Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085429.1 | NP_001078898.1 | 2 | 220 | ||
XM_005250167.3 | XP_005250224.1 | 2 | 724 | |||
GenBank mRNA | BC131697.1 | - | 2 | 263 | 40120595 |