Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284355.2 | NP_001271284.1 | 2 | 599 | ||
NM_001321766.1 | NP_001308695.1 | 2 | 599 | |||
NM_133463.3 | NP_597720.1 | 2 | 654 | |||
XM_011515150.2 | XP_011513452.1 | 2 | 599 | |||
XM_011515151.2 | XP_011513453.1 | 2 | 654 | |||
XM_017011774.1 | XP_016867263.1 | 2 | 599 | |||
XM_017011775.1 | XP_016867264.1 | 2 | 654 | |||
XM_017011776.1 | XP_016867265.1 | 2 | 654 | |||
GenBank mRNA | AB075830.1 | - | 2 | 647 | ||
AJ635357.1 | - | 1 | 286 | |||
AK092107.1 | - | 2 | 599 | |||
AK289723.1 | - | 2 | 654 | |||
BC146753.1 | - | 2 | 647 | 9007189 |