Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194291.2 | NP_919267.2 | 3, 4 | 958 | ||
XM_011516847.2 | XP_011515149.1 | 3, 4 | 952 | |||
GenBank mRNA | BC017881.1 | - | 1, 2 | 62 | ||
BC032396.1 | - | 3, 4 | 259 | 5197382 | ||
BC041379.1 | - | 3, 4 | 955 | 5275048 |