Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144966.5 | NP_659403.4 | 23 | 4682 | ||
XM_005251382.3 | XP_005251439.1 | 22 | 4930 | |||
XM_006716729.3 | XP_006716792.1 | 2 | 171 | |||
XM_017014316.1 | XP_016869805.1 | 22 | 4957 | |||
XM_017014317.1 | XP_016869806.1 | 23 | 4500 | |||
XM_017014318.1 | XP_016869807.1 | 23 | 5255 | |||
XM_017014319.1 | XP_016869808.1 | 24 | 5159 | |||
XM_017014320.1 | XP_016869809.1 | 23 | 5086 | |||
XM_017014321.1 | XP_016869810.1 | 22 | 4749 | |||
XM_017014322.1 | XP_016869811.1 | 22 | 4543 | |||
XM_017014323.1 | XP_016869812.1 | 22 | 4216 | |||
XM_017014324.1 | XP_016869813.1 | 22 | 4957 | |||
XM_017014325.1 | XP_016869814.1 | 22 | 4957 | |||
XM_017014326.1 | XP_016869815.1 | 21 | 3773 | |||
XM_017014327.1 | XP_016869816.1 | 18 | 3142 | |||
XM_017014328.1 | XP_016869817.1 | 22 | 4957 | |||
XM_017014329.1 | XP_016869818.1 | 22 | 4957 | |||
XM_017014330.1 | XP_016869819.1 | 22 | 4957 | |||
GenBank mRNA | AB160987.1 | - | 21 | 3866 | ||
BX537574.1 | - | 23 | 4683 |