Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139246.4 | NP_640339.4 | 3 | 746 | ||
XM_011518312.2 | XP_011516614.1 | 2 | 209 | |||
XM_017014342.1 | XP_016869831.1 | 3 | 392 | |||
GenBank mRNA | AK094043.1 | - | 3 | 459 | ||
AK128615.1 | - | 4 | 412 | |||
BX648493.1 | - | 3 | 385 | |||
CN298691.1 | - | 3 | 610 | |||
KU178877.1 | - | 2 | 177 | |||
KU178878.1 | - | 2 | 149 |