Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VEGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025366.2 | NP_001020537.2 | 8 | 3105 | ||
NM_001025367.2 | NP_001020538.2 | 8 | 3036 | |||
NM_001025368.2 | NP_001020539.2 | 7 | 2982 | |||
NM_001025369.2 | NP_001020540.2 | 7 | 2947 | |||
NM_001025370.2 | NP_001020541.2 | 6 | 2850 | |||
NM_001033756.2 | NP_001028928.1 | 7 | 2916 | |||
NM_001171622.1 | NP_001165093.1 | 5 | 2820 | |||
NM_001171623.1 | NP_001165094.1 | 8 | 3105 | |||
NM_001171624.1 | NP_001165095.1 | 8 | 3054 | |||
NM_001171625.1 | NP_001165096.1 | 8 | 3036 | |||
NM_001171626.1 | NP_001165097.1 | 7 | 2982 | |||
NM_001171627.1 | NP_001165098.1 | 7 | 2947 | |||
NM_001171628.1 | NP_001165099.1 | 6 | 2850 | |||
NM_001171629.1 | NP_001165100.1 | 7 | 2916 | |||
NM_001171630.1 | NP_001165101.1 | 5 | 2820 | |||
NM_001204384.1 | NP_001191313.1 | 7 | 2922 | |||
NM_001204385.1 | NP_001191314.1 | 7 | 2922 | |||
NM_001287044.1 | NP_001273973.1 | 7 | 1995 | |||
NM_001317010.1 | NP_001303939.1 | 7 | 2982 | |||
NM_003376.5 | NP_003367.4 | 8 | 3054 | |||
GenBank mRNA | AB209485.1 | - | 3 | 11616 | ||
AF022375.1 | - | 7 | 2645 | |||
AF024710.1 | - | 1 | 1368 | |||
AK125666.1 | - | 1 | 1900 | |||
BC065522.1 | - | 6 | 2277 |