Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139282.2 | NP_644811.1 | 1, 2 | 481 | ||
XM_011531281.2 | XP_011529583.1 | 2, 3 | 532 | |||
GenBank mRNA | AY099086.1 | - | 1, 2 | 481 | ||
BC069324.1 | - | 1, 2 | 456 | 7262157 | ||
BC069529.1 | - | 1, 2 | 458 | 7262169 | ||
BC103857.1 | - | 1, 2 | 440 | 40003132 | ||
BC103858.1 | - | 1, 2 | 440 | 40003136 |