Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s46157
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
17
2458
NM_152586.3
NP_689799.3
17
2556
XM_005269582.2
XP_005269639.1
16
5634
XM_011539368.2
XP_011537670.1
17
5700
XM_017015759.1
XP_016871248.1
19
5129
XM_017015760.1
XP_016871249.1
19
4031
XM_017015761.1
XP_016871250.1
19
4869
XM_017015762.1
XP_016871251.1
19
4039
XM_017015763.1
XP_016871252.1
19
3333
XM_017015764.1
XP_016871253.1
19
3396
XM_017015765.1
XP_016871254.1
20
3649
XM_017015766.1
XP_016871255.1
19
5255
XM_017015767.1
XP_016871256.1
19
4914
XM_017015768.1
XP_016871257.1
19
3085
XM_017015769.1
XP_016871258.1
19
2693
XM_017015770.1
XP_016871259.1
19
3354
XM_017015771.1
XP_016871260.1
18
5871
XM_017015772.1
XP_016871261.1
18
5877
XM_017015773.1
XP_016871262.1
17
5811
XM_017015774.1
XP_016871263.1
18
4797
XM_017015775.1
XP_016871264.1
19
4675
XM_017015776.1
XP_016871265.1
18
5180
XM_017015777.1
XP_016871266.1
18
4839
XM_017015778.1
XP_016871267.1
17
5757
XM_017015779.1
XP_016871268.1
18
5877
XM_017015780.1
XP_016871269.1
18
5877
XM_017015781.1
XP_016871270.1
17
5706
XM_017015782.1
XP_016871271.1
18
3457
XM_017015783.1
XP_016871272.1
18
4504
XM_017015784.1
XP_016871273.1
17
2450
XM_017015785.1
XP_016871274.1
17
5757
XM_017015786.1
XP_016871275.1
17
2947
GenBank mRNA
AK303633.1
-
2
149
BX649087.1
-
18
3099
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001008863, XM_008770560, CB726956.This siRNA also targets the following transcripts in Mouse: NM_030180, CK638913.
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0