Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145727.2 | NP_001139199.1 | 4 | 540 | ||
NM_001145728.2 | NP_001139200.1 | 6 | 1026 | |||
NM_001256266.1 | NP_001243195.1 | 6 | 938 | |||
NM_152590.3 | NP_689803.2 | 5 | 907 | |||
XM_011520575.1 | XP_011518877.1 | 8 | 1132 | |||
XM_011520576.2 | XP_011518878.1 | 8 | 1132 | |||
XM_011520577.2 | XP_011518879.1 | 8 | 1542 | |||
XM_011520578.2 | XP_011518880.1 | 8 | 1155 | |||
XM_011520579.2 | XP_011518881.1 | 8 | 1132 | |||
XM_011520582.1 | XP_011518884.1 | 8 | 1144 | |||
XM_011520583.1 | XP_011518885.1 | 7 | 1013 | |||
XM_011520584.1 | XP_011518886.1 | 5 | 820 | |||
XM_011520585.2 | XP_011518887.1 | 5 | 743 | |||
XM_011520587.1 | XP_011518889.1 | 5 | 516 | |||
XM_011520588.1 | XP_011518890.1 | 4 | 438 | |||
XM_017018891.1 | XP_016874380.1 | 7 | 964 | |||
XM_017018892.1 | XP_016874381.1 | 7 | 1034 | |||
XM_017018893.1 | XP_016874382.1 | 6 | 843 | |||
XM_017018894.1 | XP_016874383.1 | 8 | 1132 | |||
GenBank mRNA | AK093323.1 | - | 5 | 868 | ||
AK296816.1 | - | 4 | 532 | |||
AK302292.1 | - | 6 | 1026 | |||
BC037957.1 | - | 6 | 907 | 5744078 | ||
BP359528.1 | - | 3 | 359 |