Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178126.3 | NP_835227.1 | 7 | 876 | ||
NR_026697.1 | 6 | 845 | ||||
XM_011524440.2 | XP_011522742.1 | 7 | 629 | |||
GenBank mRNA | AK091125.1 | - | 6 | 1221 | ||
AK225797.1 | - | 7 | 824 | |||
AK294791.1 | - | 6 | 782 | |||
AK308288.1 | - | 7 | 619 | |||
BC049370.1 | - | 7 | 833 | 5263052 | ||
BX647175.1 | - | 7 | 767 |