Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153357.1 | NP_699188.1 | 3 | 900 | ||
XM_005256488.3 | XP_005256545.2 | 4 | 1133 | |||
XM_017024281.1 | XP_016879770.1 | 4 | 1133 | |||
GenBank mRNA | AK074674.1 | - | 3 | 900 | ||
BC093860.1 | - | 3 | 583 | 7939705 | ||
BC093886.1 | - | 3 | 583 | 7939731 |