Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300979.1 | NP_001287908.1 | 6 | 840 | ||
NM_001322911.1 | NP_001309840.1 | 5 | 713 | |||
NM_001322912.1 | NP_001309841.1 | 5 | 765 | |||
NM_001322913.1 | NP_001309842.1 | 6 | 844 | |||
NM_001322914.1 | NP_001309843.1 | 6 | 1016 | |||
NM_001322915.1 | NP_001309844.1 | 7 | 912 | |||
NM_001322916.1 | NP_001309845.1 | 6 | 788 | |||
NM_001322917.1 | NP_001309846.1 | 6 | 792 | |||
NM_001322918.1 | NP_001309847.1 | 7 | 916 | |||
NM_001322919.1 | NP_001309848.1 | 6 | 1012 | |||
NM_001322920.1 | NP_001309849.1 | 7 | 964 | |||
NM_152603.4 | NP_689816.2 | 5 | 717 | |||
XM_011526584.2 | XP_011524886.1 | 4 | 644 | |||
XM_011526585.2 | XP_011524887.1 | 4 | 644 | |||
XM_017026416.1 | XP_016881905.1 | 5 | 976 | |||
XM_017026417.1 | XP_016881906.1 | 8 | 2059 | |||
XM_017026418.1 | XP_016881907.1 | 6 | 919 | |||
XM_017026419.1 | XP_016881908.1 | 5 | 975 | |||
XM_017026420.1 | XP_016881909.1 | 6 | 1828 | |||
XM_017026421.1 | XP_016881910.1 | 5 | 939 | |||
GenBank mRNA | AK093034.1 | - | 5 | 649 | ||
AK126691.1 | - | 3 | 1739 | |||
AK298556.1 | - | 5 | 707 | |||
AK303326.1 | - | 6 | 824 | |||
AK313647.1 | - | 5 | 713 | |||
BC033849.2 | - | 4 | 691 | 4472579 | ||
BX640706.1 | - | 6 | 829 |