Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320257.1 | NP_001307186.1 | 4 | 579 | ||
NM_198460.2 | NP_940862.2 | 6 | 920 | |||
XM_011540835.2 | XP_011539137.1 | 6 | 749 | |||
GenBank mRNA | AK131356.1 | - | 6 | 904 | ||
AK290303.1 | - | 6 | 740 | |||
AK299408.1 | - | 5 | 555 | |||
AK299466.1 | - | 4 | 396 | |||
AK303987.1 | - | 6 | 664 | |||
AK315834.1 | - | 6 | 738 | |||
BC131713.1 | - | 6 | 698 | 40122722 | ||
BX537949.1 | - | 6 | 739 | |||
BX647907.1 | - | 6 | 737 |