Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 5 | 548 | ||
NM_001300858.1 | NP_001287787.1 | 6 | 759 | |||
NM_144977.4 | NP_659414.2 | 5 | 548 | |||
NR_125340.1 | 5 | 548 | ||||
XM_005244931.2 | XP_005244988.1 | 5 | 548 | |||
XM_006711192.2 | XP_006711255.1 | 6 | 458 | |||
XM_006711193.3 | XP_006711256.1 | 6 | 590 | |||
XM_011509246.2 | XP_011507548.1 | 5 | 409 | |||
XM_011509247.1 | XP_011507549.1 | 5 | 409 | |||
XM_011509248.2 | XP_011507550.1 | 5 | 342 | |||
XM_011509249.2 | XP_011507551.1 | 6 | 647 | |||
XM_011509251.2 | XP_011507553.1 | 5 | 411 | |||
XM_017000470.1 | XP_016855959.1 | 6 | 536 | |||
XM_017000471.1 | XP_016855960.1 | 6 | 458 | |||
XM_017000472.1 | XP_016855961.1 | 3 | 128 | |||
GenBank mRNA | AK091207.1 | - | 5 | 362 | ||
BC022561.1 | - | 6 | 461 | 4793412 | ||
BC063877.1 | - | 5 | 380 | 30377843 | ||
DA495773.1 | - | 5 | 548 |