Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181712.4 | NP_859063.3 | 3 | 1546 | ||
XM_011540843.2 | XP_011539145.1 | 3 | 1431 | |||
XM_011540845.2 | XP_011539147.1 | 3 | 1497 | |||
XM_017000483.1 | XP_016855972.1 | 3 | 1496 | |||
GenBank mRNA | AB284127.1 | - | 3 | 1546 | ||
AL832644.2 | - | 1 | 298 | |||
BC060866.1 | - | 3 | 1546 | 30344114 |