Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RASSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270391.1 | NP_001257320.1 | 9 | 1139 | ||
NM_001270392.1 | NP_001257321.1 | 9 | 1019 | |||
NM_177532.4 | NP_803876.1 | 10 | 1241 | |||
NM_201431.2 | NP_958834.1 | 10 | 1151 | |||
XM_011531687.2 | XP_011529989.1 | 10 | 1143 | |||
XM_011531688.1 | XP_011529990.1 | 11 | 1217 | |||
XM_011531689.2 | XP_011529991.1 | 9 | 1041 | |||
XM_011531690.2 | XP_011529992.1 | 9 | 3777 | |||
XM_011531691.1 | XP_011529993.1 | 8 | 869 | |||
XM_017007837.1 | XP_016863326.1 | 7 | 3121 | |||
GenBank mRNA | AK126346.1 | - | 10 | 1143 | ||
AY217664.1 | - | 10 | 1055 | |||
AY217665.1 | - | 9 | 888 | |||
BC058835.1 | - | 9 | 982 | 6668006 | ||
CR749283.1 | - | 10 | 1241 |