Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133468.4 | NP_597725.1 | 13 | 1495 | ||
XM_017011800.1 | XP_016867289.1 | 11 | 1055 | |||
GenBank mRNA | AB075845.1 | - | 9 | 762 | ||
AK289963.1 | - | 12 | 1235 | |||
AY324650.1 | - | 13 | 1494 | |||
BC060868.1 | - | 12 | 1172 | 30345290 |