Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STKLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153710.4 | NP_714921.4 | 12 | 1190 | ||
NR_103997.1 | 10 | 2039 | ||||
GenBank mRNA | AK131533.1 | - | 9 | 2588 | ||
AK131545.1 | - | 10 | 1991 | |||
BC036504.2 | - | 12 | 1190 | 5265506 |