Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133474.3 | NP_597731.2 | 3 | 572 | ||
GenBank mRNA | AK127067.1 | - | 1 | 128 | ||
AK128499.1 | - | 1 | 401 | |||
AL834155.1 | - | 376 | ||||
DA309597.1 | - | 3 | 555 |
Gene Symbol: ZNF732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 548 | ||
GenBank mRNA | AK302099.1 | - | 3 | 548 |