Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 16 | 1493 | ||
XM_011527756.2 | XP_011526058.1 | 18 | 1598 | |||
XM_011527757.2 | XP_011526059.1 | 16 | 1651 | |||
XM_011527758.2 | XP_011526060.1 | 17 | 1511 | |||
XM_011527761.1 | XP_011526063.1 | 19 | 1668 | |||
XM_011527764.2 | XP_011526066.1 | 15 | 1564 | |||
XM_011527765.2 | XP_011526067.1 | 16 | 1412 | |||
XM_011527768.2 | XP_011526070.1 | 4 | 369 | |||
XM_017026438.1 | XP_016881927.1 | 19 | 1659 | |||
XM_017026439.1 | XP_016881928.1 | 18 | 1581 | |||
XM_017026440.1 | XP_016881929.1 | 17 | 1554 | |||
XM_017026441.1 | XP_016881930.1 | 7 | 696 | |||
GenBank mRNA | AB075861.1 | - | 14 | 1190 | ||
BC028236.1 | - | 14 | 1135 | |||
BX359277.2 | - | 4 | 242 |