Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 20 | 3476 | ||
XM_011527756.2 | XP_011526058.1 | 22 | 3581 | |||
XM_011527757.2 | XP_011526059.1 | 20 | 3634 | |||
XM_011527758.2 | XP_011526060.1 | 21 | 3494 | |||
XM_011527761.1 | XP_011526063.1 | 23 | 3651 | |||
XM_011527764.2 | XP_011526066.1 | 19 | 3547 | |||
XM_011527765.2 | XP_011526067.1 | 20 | 3395 | |||
XM_011527768.2 | XP_011526070.1 | 8 | 2352 | |||
XM_017026438.1 | XP_016881927.1 | 23 | 3642 | |||
XM_017026439.1 | XP_016881928.1 | 22 | 3564 | |||
XM_017026440.1 | XP_016881929.1 | 21 | 3537 | |||
XM_017026441.1 | XP_016881930.1 | 11 | 2679 |