Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015338.5 | NP_056153.2 | 12, 13 | 2157 | ||
XM_006723727.3 | XP_006723790.1 | 11, 12 | 2436 | |||
XM_006723728.3 | XP_006723791.1 | 12, 13 | 1766 | |||
XM_006723730.3 | XP_006723793.1 | 12, 13 | 1962 | |||
XM_006723732.2 | XP_006723795.1 | 11, 12 | 1785 | |||
XM_006723733.1 | XP_006723796.1 | 5, 6 | 1791 | |||
XM_011528648.2 | XP_011526950.1 | 12, 13 | 2081 | |||
XM_011528652.2 | XP_011526954.1 | 13, 14 | 3099 | |||
XM_017027704.1 | XP_016883193.1 | 12, 13 | 3222 | |||
XM_017027705.1 | XP_016883194.1 | 12, 13 | 2235 | |||
XM_017027706.1 | XP_016883195.1 | 11, 12 | 2286 | |||
GenBank mRNA | AB023195.2 | - | 6, 7 | 1207 | ||
AB084280.1 | - | 11, 12 | 1710 | |||
AJ438952.2 | - | 12, 13 | 1983 | |||
AK122923.1 | - | 11, 12 | 1785 | |||
AL117518.1 | - | 5, 6 | 1127 | |||
BC137278.1 | - | 13, 14 | 1920 | 9021275 | ||
BC137280.1 | - | 13, 14 | 1920 | 9021277 |