Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205849.3 | NP_995321.1 | 7 | 796 | ||
XM_011545464.2 | XP_011543766.1 | 14 | 1369 | |||
GenBank mRNA | AF494344.1 | - | 7 | 795 | ||
AK097501.1 | - | 6 | 740 | |||
AK128470.1 | - | 7 | 3157 | |||
BC120955.1 | - | 6 | 495 | 40115245 | ||
BC120956.1 | - | 6 | 495 | 40115246 |