Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243168.1 | NP_001230097.1 | 4 | 1080 | ||
NM_001243169.1 | NP_001230098.1 | 5 | 960 | |||
NM_001243170.1 | NP_001230099.1 | 3 | 714 | |||
NM_138296.2 | NP_612153.2 | 4 | 1035 | |||
GenBank mRNA | AK130819.1 | - | 1392 | |||
AK303523.1 | - | 4 | 510 | |||
BC100771.1 | - | 4 | 977 | 40000277 | ||
BC100772.1 | - | 4 | 977 | 40000280 | ||
BC100773.2 | - | 4 | 977 | 40000282 | ||
BC100774.2 | - | 4 | 977 | 40000286 | ||
BC153829.1 | - | 4 | 977 | 40000274 | ||
U36759.1 | - | 4 | 1035 | |||
U38996.1 | - | 4 | 919 |