Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182832.2 | NP_878252.2 | 1 | 6293 | ||
GenBank mRNA | AF269287.1 | - | 1 | 2542 | ||
AK027868.1 | - | 1 | 3955 | |||
L13197.1 | - | 2 | 1925 |