Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 5, 6 | 779 | ||
NM_001322020.1 | NP_001308949.1 | 5, 6 | 779 | |||
NM_004544.3 | NP_004535.1 | 5, 6 | 779 | |||
XM_011511228.2 | XP_011509530.1 | 5, 6 | 766 | |||
GenBank mRNA | AF087661.1 | - | 5, 6 | 699 | ||
AF087854.1 | - | 5, 6 | 718 | |||
AK290778.1 | - | 5, 6 | 701 | |||
BC003417.1 | - | 5, 6 | 713 | 3451514 |