Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286245.1 | NP_001273174.1 | 6 | 1886 | ||
NM_001286246.1 | NP_001273175.1 | 6 | 1905 | |||
NM_178502.3 | NP_848597.1 | 7 | 2066 | |||
XM_005268697.1 | XP_005268754.1 | 7 | 2569 | |||
XM_005268698.1 | XP_005268755.1 | 7 | 2342 | |||
XM_005268700.1 | XP_005268757.1 | 7 | 2222 | |||
XM_005268703.1 | XP_005268760.1 | 7 | 2241 | |||
XM_011538022.2 | XP_011536324.1 | 6 | 2444 | |||
GenBank mRNA | AK092085.1 | - | 7 | 1867 | ||
AK094385.1 | - | 6 | 1675 | |||
AK128752.1 | - | 5 | 3034 |