Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143912.1 | NP_001137384.1 | 8 | 918 | ||
NM_001143913.1 | NP_001137385.1 | 7 | 831 | |||
NM_001143914.1 | NP_001137386.1 | 6 | 729 | |||
NM_001143915.1 | NP_001137387.1 | 5 | 576 | |||
NM_152660.2 | NP_689873.1 | 7 | 816 | |||
GenBank mRNA | AK098318.1 | - | 6 | 680 | ||
AK290120.1 | - | 6 | 724 | |||
AK301672.1 | - | 5 | 559 | |||
AK310165.1 | - | 7 | 767 | |||
AL035307.1 | - | 8 | 840 | |||
AL035309.1 | - | 7 | 753 | |||
BC025768.1 | - | 7 | 816 | 5211010 |