Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175852.3 | NP_787048.1 | 6, 7 | 1030 | ||
XM_011540931.2 | XP_011539233.1 | 5, 6 | 953 | |||
XM_011540932.2 | XP_011539234.1 | 4, 5 | 790 | |||
XM_017000561.1 | XP_016856050.1 | 5, 6 | 1422 | |||
XM_017000562.1 | XP_016856051.1 | 5, 6 | 1323 | |||
XM_017000563.1 | XP_016856052.1 | 6, 7 | 1204 | |||
XM_017000564.1 | XP_016856053.1 | 5, 6 | 884 | |||
XM_017000565.1 | XP_016856054.1 | 5, 6 | 978 | |||
GenBank mRNA | AF516206.1 | - | 5, 6 | 1043 | ||
AL832338.1 | - | 6, 7 | 1067 | |||
AL832636.2 | - | 6, 7 | 1089 | |||
AL832637.2 | - | 5, 6 | 1246 | |||
BC080578.1 | - | 5, 6 | 1007 | 6598234 | ||
BC103823.1 | - | 5, 6 | 995 | 40001224 | ||
BC103824.1 | - | 5, 6 | 995 | 40001225 |