Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012970.2 | NP_001012988.1 | 3 | 241 | ||
NM_001276348.1 | NP_001263277.1 | 3 | 241 | |||
NM_001276349.1 | NP_001263278.1 | 3 | 241 | |||
XM_011544124.2 | XP_011542426.1 | 4 | 1083 | |||
XM_011544125.2 | XP_011542427.1 | 3 | 809 | |||
XM_011544128.2 | XP_011542430.1 | 3 | 1320 | |||
XM_011544129.2 | XP_011542431.1 | 4 | 1090 | |||
XM_011544130.2 | XP_011542432.1 | 4 | 1088 | |||
XM_017000571.1 | XP_016856060.1 | 4 | 1089 | |||
XM_017000572.1 | XP_016856061.1 | 4 | 1597 | |||
GenBank mRNA | BC054479.1 | - | 3 | 245 | ||
BX099227.1 | - | 3 | 227 | |||
HY055397.1 | - | 3 | 197 |