Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 5 | 888 | ||
NM_001174151.1 | NP_001167622.1 | 4 | 817 | |||
NM_001321328.1 | NP_001308257.1 | 6 | 1010 | |||
NM_144996.3 | NP_659433.2 | 3 | 567 | |||
NM_182896.2 | NP_878899.1 | 5 | 888 | |||
NR_033427.1 | 4 | 643 | ||||
NR_135621.1 | 4 | 634 | ||||
XM_006713532.3 | XP_006713595.1 | 6 | 1007 | |||
XM_011512532.2 | XP_011510834.1 | 6 | 916 | |||
XM_011512533.2 | XP_011510835.1 | 7 | 724 | |||
XM_011512534.2 | XP_011510836.1 | 7 | 1042 | |||
XM_011512535.2 | XP_011510837.1 | 6 | 901 | |||
XM_017005853.1 | XP_016861342.1 | 6 | 918 | |||
GenBank mRNA | AK296994.1 | - | 5 | 866 | ||
AK299284.1 | - | 4 | 627 | |||
AL713789.1 | - | 4 | 759 | |||
BC015439.1 | - | 3 | 527 | |||
BC094725.1 | - | 3 | 562 | 4818884 | ||
BC104035.1 | - | 5 | 878 | 40026398 | ||
BC104036.1 | - | 5 | 878 | 40026399 | ||
CR936756.1 | - | 4 | 522 | |||
CR936763.1 | - | 6 | 962 |