Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPL22L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099645.1 | NP_001093115.1 | 3 | 250 | ||
NM_001320451.1 | NP_001307380.1 | 3 | 247 | |||
NR_135259.1 | 3 | 317 | ||||
GenBank mRNA | AL110170.1 | - | 3 | 244 | ||
BC062731.1 | - | 3 | 193 | |||
BC107708.1 | - | 3 | 175 | 6618915 | ||
BM451642.1 | - | 3 | 262 | |||
BU540139.1 | - | 3 | 189 |