Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 2 | 595 | ||
NM_001291833.1 | NP_001278762.1 | 2 | 589 | |||
NM_020790.1 | NP_065841.1 | 2 | 592 | |||
NM_152673.3 | NP_689886.3 | 2 | 595 | |||
GenBank mRNA | AB037780.1 | - | 1 | 2233 | ||
AB098731.1 | - | 1 | 846 | |||
AK027314.1 | - | 2 | 499 | |||
AK298316.1 | - | 2 | 717 | |||
AK311504.1 | - | 2 | 593 | |||
AY358449.1 | - | 2 | 528 | |||
BC029267.1 | - | 2 | 503 | 3851952 | ||
BC044243.1 | - | 2 | 482 | 5754328 | ||
BM721408.1 | - | 2 | 423 |