Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194296.1 | NP_919272.1 | 2 | 164 | ||
XM_005271916.4 | XP_005271973.1 | 2 | 281 | |||
XM_011543252.2 | XP_011541554.1 | 2 | 165 | |||
XM_011543253.2 | XP_011541555.1 | 2 | 143 | |||
XM_011543254.2 | XP_011541556.1 | 2 | 214 | |||
GenBank mRNA | AK098740.1 | - | 2 | 165 | ||
BC040037.1 | - | 2 | 144 | |||
BC050563.1 | - | 2 | 144 | 5751687 | ||
BM785501.1 | - | 2 | 165 |