Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC125 SIRNA Assays ›
Gene Symbol:
CCDC125
Gene Name:
coiled-coil domain containing 125
Gene Aliases:
KENAE
Chromosome Location:
Chr.5: 69273087 - 69333069 on Build GRCh38
UniGene ID:
Hs.654850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47436
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC125
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297696.1
NP_001284625.1
11
1205
NM_001297697.1
NP_001284626.1
13
1261
NM_176816.4
NP_789786.2
12
1422
XM_005248458.4
XP_005248515.1
12
1659
XM_005248459.4
XP_005248516.1
12
1461
XM_005248461.3
XP_005248518.1
11
1148
XM_005248463.3
XP_005248520.1
10
1016
XM_005248464.3
XP_005248521.1
11
1289
XM_006714569.3
XP_006714632.1
12
1449
XM_006714570.3
XP_006714633.1
12
1233
XM_011543255.2
XP_011541557.1
12
1439
XM_011543256.2
XP_011541558.1
12
1440
XM_011543257.2
XP_011541559.1
11
1356
XM_011543258.2
XP_011541560.1
12
1464
XM_011543259.2
XP_011541561.1
12
1464
XM_017009206.1
XP_016864695.1
12
1654
XM_017009207.1
XP_016864696.1
12
1449
XM_017009208.1
XP_016864697.1
10
1288
XM_017009209.1
XP_016864698.1
11
1286
XM_017009210.1
XP_016864699.1
9
1209
XM_017009211.1
XP_016864700.1
6
691
GenBank mRNA
AB024691.1
-
12
1422
AB024692.1
-
11
1125
AK299871.1
-
13
1261
BC108715.1
-
10
1181
BC113749.1
-
6
674
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: BC098689.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0