Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313994.1 | NP_001300923.1 | 13 | 1593 | ||
NM_145004.6 | NP_659441.3 | 16 | 1991 | |||
XM_006716298.2 | XP_006716361.1 | 16 | 1785 | |||
XM_006716299.1 | XP_006716362.1 | 14 | 1467 | |||
XM_011544433.1 | XP_011542735.1 | 16 | 1785 | |||
XM_011544434.1 | XP_011542736.1 | 14 | 1566 | |||
XM_017013183.1 | XP_016868672.1 | 15 | 1716 | |||
XM_017013184.1 | XP_016868673.1 | 16 | 1785 | |||
XM_017013185.1 | XP_016868674.1 | 15 | 1791 | |||
XM_017013186.1 | XP_016868675.1 | 12 | 1395 | |||
XM_017013187.1 | XP_016868676.1 | 8 | 931 | |||
XM_017013188.1 | XP_016868677.1 | 7 | 900 | |||
GenBank mRNA | AK131038.1 | - | 8 | 960 | ||
AK313864.1 | - | 16 | 1812 | |||
AY358739.1 | - | 16 | 1871 | |||
BC026085.2 | - | 16 | 1775 | 4838404 | ||
BC026169.2 | - | 16 | 1802 | 4829390 | ||
BC028702.2 | - | 16 | 1785 | 4829385 | ||
BC030014.2 | - | 16 | 1778 | 4830659 | ||
BC030698.2 | - | 16 | 1793 | 4822988 | ||
BC034975.2 | - | 16 | 1786 | 4829806 |